2-(Methylsulfonyl)phenol ≥95%

Supplier: Matrix Scientific

044719-500MG 044719-1G 044719-5G
101867-252EA 146.15 USD
101867-252 101867-254 101867-256
2-(Methylsulfonyl)phenol ≥95%
2-(Methylsulphonyl)phenol

Hazard: Irritant

Formula: C₇H₈O₃S
Melting Pt: 88…90 °C
MDL Number: MFCD00127516
CAS Number: 27489-33-4

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR