4-[(Dimethylamino)methylene]-2,4-dihydro-3H-pyrazol-3-one ≥95%

Supplier: Matrix Scientific

044718-500MG 044718-1G 044718-5G
101867-246EA 375.38 USD
101867-246 101867-248 101867-250
4-[(Dimethylamino)methylene]-2,4-dihydro-3H-pyrazol-3-one ≥95%
4-[(Dimethylamino)methylene]-2,4-dihydro-3H-pyrazol-3-one

Hazard: Irritant

Formula: C₆H₉N₃O
MW: 139.16 g/mol
Melting Pt: 243…245 °C
MDL Number: MFCD00138484
CAS Number: 821016-54-0

Order Now

Specification Test Results

Melting point 243-245°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR