2-(2-Bromobenzoyl)-3-(dimethylamino)acrylonitrile ≥95%

Supplier: Matrix Scientific

044712-500MG 044712-1G 044712-5G
101867-222EA 278.46 USD
101867-222 101867-224 101867-226
2-(2-Bromobenzoyl)-3-(dimethylamino)acrylonitrile ≥95%
2-(2-Bromobenzoyl)-3-(dimethylamino)acrylonitrile

Hazard: Irritant

Formula: C₁₂H₁₁BrN₂O
Melting Pt: 88…90 °C
MDL Number: MFCD08276956
CAS Number: 886361-83-7

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR