N'-Hydroxy-2-pyrazinecarboximidamide ≥95%

Supplier: Matrix Scientific

Synonyms: N'-Hydroxypyrazine-2-carboximidamide, Pyrazine-2-amidoxime

044710-500MG 044710-1G 044710-5G
101867-212EA 541.54 USD
101867-212 101867-214 101867-216
N'-Hydroxy-2-pyrazinecarboximidamide ≥95%
N'-Hydroxy-2-pyrazinecarboximidamide

Hazard: Irritant

Formula: C₅H₆N₄O
MW: 138.13 g/mol
Melting Pt: 183…185 °C
MDL Number: MFCD02258397
CAS Number: 51285-05-3

Order Now

Specification Test Results

Melting point 183-185°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR