Ethyl-1-(2,6-dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

044709-500MG 044709-1G 044709-5G
101867-206EA 375.38 USD
101867-206 101867-208 101867-210
Ethyl-1-(2,6-dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-(2,6-dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄Cl₂N₂O₂
Melting Pt: 88…90 °C
MDL Number: MFCD08056634
CAS Number: 1006441-21-9

Order Now

Specification Test Results

Melting point 88-90°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR