N'-Hydroxy-2-thiophenecarboximidamide ≥95%

Supplier: Matrix Scientific

Synonyms: N-Hydroxythiophene-2-carboximidamide

044707-1G 044707-5G
101867-202EA 129.23 USD
101867-202 101867-204
N'-Hydroxy-2-thiophenecarboximidamide ≥95%
N'-Hydroxy-2-thiophenecarboximidamide

Hazard: Irritant

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Melting Pt: 94…97 °C
MDL Number: MFCD00173698
CAS Number: 53370-51-7

Order Now

Specification Test Results

Melting point 94-97°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR