[1-(4-Chlorophenyl)-1H-1,2,3-triazol-4-yl]methanamine ≥95%

Supplier: Matrix Scientific

044703-500MG 044703-1G
101867-188EA 747.69 USD
101867-188 101867-190
[1-(4-Chlorophenyl)-1H-1,2,3-triazol-4-yl]methanamine ≥95%
[1-(4-Chlorophenyl)-1H-1,2,3-triazol-4-yl]methanamine

Hazard: Irritant

Formula: C₉H₉ClN₄
Melting Pt: 99…101 °C
MDL Number: MFCD08056633
CAS Number: 886361-79-1

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR