2-Amino-1-(4-fluorophenyl)-1-ethanol ≥95%

Supplier: Matrix Scientific

044702-500MG 044702-1G
101867-184EA 747.69 USD
101867-184 101867-186
2-Amino-1-(4-fluorophenyl)-1-ethanol ≥95%
2-Amino-1-(4-fluorophenyl)-1-ethanol

Hazard: Irritant

Formula: C₈H₁₀FNO
Melting Pt: 60…63 °C
MDL Number: MFCD08056632
CAS Number: 456-05-3

Order Now

Specification Test Results

Melting point 60-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR