5-Phenylthiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Phenyl-2-thiophenecarboxylic acid, 5-Phenyl-2-thenoic acid

044701-1G 044701-250MG 044701-5G
101867-182EA 638.46 USD
101867-182 102663-718 102663-720
5-Phenylthiophene-2-carboxylic acid ≥95%
5-Phenylthiophene-2-carboxylic acid

Hazard: Irritant

Formula: C11H8O2S
Melting Pt: 184…187 °C
MDL Number: MFCD01313432
CAS Number: 19163-24-7

Order Now

Specification Test Results

Melting point 184-187°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR