5-(3-Nitrophenyl)-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

044700-500MG 044700-1G 044700-5G
101867-174EA 556.92 USD
101867-174 101867-176 101867-178
5-(3-Nitrophenyl)-2-thiophenecarboxylic acid ≥95%
5-(3-Nitrophenyl)-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₇NO₄S
Melting Pt: 264…266 °C
MDL Number: MFCD06803028
CAS Number: 341015-21-2

Order Now

Specification Test Results

Melting point 264-266°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR