Ethyl-5-phenyl-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

044699-500MG 044699-1G
101867-170EA 747.69 USD
101867-170 101867-172
Ethyl-5-phenyl-2-thiophenecarboxylate ≥95%
Ethyl-5-phenyl-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₂O₂S
Melting Pt: 31…33 °C
MDL Number: MFCD01860759
CAS Number: 19282-39-4

Order Now

Specification Test Results

Melting point 31-33°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR