Ethyl-5-(3-nitrophenyl)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

044698-500MG 044698-1G 044698-5G
101867-164EA 375.38 USD
101867-164 101867-166 101867-168
Ethyl-5-(3-nitrophenyl)-2-thiophenecarboxylate ≥95%
Ethyl-5-(3-nitrophenyl)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₁NO₄S
Melting Pt: 88…91 °C
MDL Number: MFCD08056631
CAS Number: 886361-77-9

Order Now

Specification Test Results

Melting point 88-91°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR