Ethyl-5-(4-fluorophenyl)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

044696-500MG 044696-1G 044696-5G
101867-154EA 556.92 USD
101867-154 101867-156 101867-158
Ethyl-5-(4-fluorophenyl)-2-thiophenecarboxylate ≥95%
Ethyl-5-(4-fluorophenyl)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₁FO₂S
Melting Pt: 72…74 °C
MDL Number: MFCD08056630
CAS Number: 848095-02-3

Order Now

Specification Test Results

Melting point 72-74°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR