2-{2-[4-(Trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid ≥95%

Supplier: Matrix Scientific

044693-500MG 044693-1G 044693-5G
101867-136EA 556.92 USD
101867-136 101867-138 101867-140
2-{2-[4-(Trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid ≥95%
2-{2-[4-(Trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid

Hazard: Irritant

Formula: C₁₂H₈F₃NO₂S
Melting Pt: 134…136 °C
MDL Number: MFCD08056629
CAS Number: 17969-63-0

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR