2-Amino-4'-fluoroacetophenone hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-1-(4-fluorophenyl)ethanone hydrochloride, 2-Amino-4'-fluoroacetophenone Hydrochloride, 4-Fluorophenacylamine hydrochloride
044692-1G
044692-5G
101867-132EA
186.15
USD
101867-132
101867-134
2-Amino-4'-fluoroacetophenone hydrochloride ≥95%
2-Amino-4'-fluoroacetophenone hydrochloride
Hazard: Irritant
Formula:
C₈H₉ClFNO MW: 189.6146 g/mol Melting Pt: 238-242°C |
MDL Number:
MFCD00193059 CAS Number: 456-00-8 |
Specification Test Results
Melting point | 238-242°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...