Methyl-4-(3-nitro-2-pyridinyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(3-nitropyridin-2-yl)benzoate

044691-500MG 044691-5G
101867-124EA 278.46 USD
101867-124 101867-128
Methyl-4-(3-nitro-2-pyridinyl)benzenecarboxylate ≥95%
Methyl-4-(3-nitro-2-pyridinyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₀N₂O₄
MW: 258.24 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD08443945
CAS Number: 937601-36-0

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR