5-(3-Thienyl)-2-furaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Thiophen-3-yl)furan-2-carbaldehyde

044689-500MG 044689-1G
101867-116EA 747.69 USD
101867-116 101867-118
5-(3-Thienyl)-2-furaldehyde ≥95%
5-(3-Thienyl)-2-furaldehyde

Hazard: Irritant

Formula: C₉H₆O₂S
MW: 178.21 g/mol
Melting Pt: 57…59 °C
MDL Number: MFCD06740284
CAS Number: 886361-73-5

Order Now

Specification Test Results

Melting point 57-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR