2-Fluoro-5-(3-thienyl)benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

044688-500MG 044688-1G 044688-5G
101867-110EA 556.92 USD
101867-110 101867-112 101867-114
2-Fluoro-5-(3-thienyl)benzenecarbonitrile ≥95%
2-Fluoro-5-(3-thienyl)benzenecarbonitrile

Hazard: Irritant

Formula: C₁₁H₆FNS
Melting Pt: 85…88 °C
MDL Number: MFCD07698593
CAS Number: 886361-71-3

Order Now

Specification Test Results

Melting point 85-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR