2-Nitro-5-piperidinoaniline ≥95%

Supplier: Matrix Scientific

044686-500MG 044686-1G 044686-5G
101867-098EA 181.54 USD
101867-098 101867-100 101867-102
2-Nitro-5-piperidinoaniline ≥95%
2-Nitro-5-piperidinoaniline

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
Melting Pt: 102…104 °C
MDL Number: MFCD00545948
CAS Number: 54997-99-8

Order Now

Specification Test Results

Melting point 102-104°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR