2-Nitro-5-(1-pyrrolidinyl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Nitro-5-(pyrrolidin-1-yl)aniline

044685-500MG 044685-1G 044685-5G
101867-092EA 181.54 USD
101867-092 101867-094 101867-096
2-Nitro-5-(1-pyrrolidinyl)aniline ≥95%
2-Nitro-5-(1-pyrrolidinyl)aniline

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₂
MW: 207.23 g/mol
Melting Pt: 186…188 °C
MDL Number: MFCD00545947
CAS Number: 289913-98-0

Order Now

Specification Test Results

Melting point 186-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR