5-Phenyl-4-(1H-1,2,4-triazol-1-yl)-1,3-cyclohexanedione ≥95%

Supplier: Matrix Scientific

044678-500MG 044678-1G 044678-5G
101867-058EA 556.92 USD
101867-058 101867-060 101867-062
5-Phenyl-4-(1H-1,2,4-triazol-1-yl)-1,3-cyclohexanedione ≥95%
5-Phenyl-4-(1H-1,2,4-triazol-1-yl)-1,3-cyclohexanedione

Hazard: Irritant

Formula: C₁₄H₁₃N₃O₂
MW: 255.28 g/mol
Melting Pt: 221…223 °C
MDL Number: MFCD06411615
CAS Number: 886361-84-8

Order Now

Specification Test Results

Melting point 221-223°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR