1-{1-[(6-Chloro-3-pyridinyl)methyl]-1H-indol-3-yl}-1-ethanone ≥95%
Supplier: Matrix Scientific
044677-500MG
044677-1G
044677-5G
101867-052EA
278.46
USD
101867-052
101867-054
101867-056
1-{1-[(6-Chloro-3-pyridinyl)methyl]-1H-indol-3-yl}-1-ethanone ≥95%
1-{1-[(6-Chloro-3-pyridinyl)methyl]-1H-indol-3-yl}-1-ethanone
Hazard: Irritant
Formula:
C₁₆H₁₃ClN₂O MW: 284.75 g/mol Melting Pt: 146…148 °C |
MDL Number:
MFCD06411614 CAS Number: 886361-82-6 |
Specification Test Results
Melting point | 146-148°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...