Methyl-2-(4-formylphenoxy)-2-phenylacetate ≥95%
Supplier: Matrix Scientific
044674-500MG
044674-1G
044674-5G
101867-034EA
278.46
USD
101867-034
101867-036
101867-038
Methyl-2-(4-formylphenoxy)-2-phenylacetate ≥95%
Methyl-2-(4-formylphenoxy)-2-phenylacetate
Hazard: Irritant
Formula:
C₁₆H₁₄O₄ MW: 270.29 g/mol Melting Pt: 88…89 °C |
MDL Number:
MFCD06200973 CAS Number: 692754-44-2 |
Specification Test Results
Melting point | 88-89°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...