1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

044672-500MG 044672-1G 044672-5G
101867-022EA 375.38 USD
101867-022 101867-024 101867-026
1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
1-(3-Bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₁BrN₂O₂
Melting Pt: 219…219 °C
MDL Number: MFCD06200971
CAS Number: 959582-69-5

Order Now

Specification Test Results

Melting point 219-219°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR