5-(4-Benzylpiperazino)-2-nitroaniline ≥95%
Supplier: Matrix Scientific
044665-500MG
044665-1G
044665-5G
101866-998EA
132.31
USD
101866-998
101867-000
101867-002
5-(4-Benzylpiperazino)-2-nitroaniline ≥95%
5-(4-Benzylpiperazino)-2-nitroaniline
Hazard: Irritant
Formula:
C₁₇H₂₀N₄O₂ Melting Pt: 160 °C |
MDL Number:
MFCD06200970 CAS Number: 23470-43-1 |
Specification Test Results
Melting point | 160°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...