6-Phenyl-3-pyridinamine ≥95%

Supplier: Matrix Scientific

044662-500MG 044662-1G 044662-5G
101866-980EA 100 USD
101866-980 101866-982 101866-984
6-Phenyl-3-pyridinamine ≥95%
6-Phenyl-3-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₀N₂
MW: 170.22 g/mol
Melting Pt: 104…106 °C
MDL Number: MFCD04114197
CAS Number: 126370-67-0

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR