5-Amino-2-(4-methoxyphenyl)pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
6-(4-Methoxyphenyl)pyridin-3-amine
044660-500MG
044660-1G
044660-5G
101866-968EA
556.92
USD
101866-968
101866-970
101866-972
5-Amino-2-(4-methoxyphenyl)pyridine ≥95%
5-Amino-2-(4-methoxyphenyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O Melting Pt: 110-112°C |
MDL Number:
MFCD05864840 CAS Number: 52057-98-4 |
Specification Test Results
Melting point | 110-112°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...