5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole ≥95%

Supplier: Matrix Scientific

044652-500MG 044652-1G 044652-5G
101866-920EA 181.54 USD
101866-920 101866-922 101866-924
5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole ≥95%
5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole

Hazard: Irritant

Formula: C₁₆H₁₆BrNO₂
MW: 334.22 g/mol
Melting Pt: 179…181 °C
MDL Number: MFCD06411613
CAS Number: 282546-99-0

Order Now

Specification Test Results

Melting point 179-181°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR