1,4-Bis(2-chloroethoxy)benzene ≥95%
Supplier: Matrix Scientific
044651-500MG
044651-1G
101866-916EA
278.46
USD
101866-916
101866-918
1,4-Bis(2-chloroethoxy)benzene ≥95%
1,4-Bis(2-chloroethoxy)benzene
Hazard: Irritant
Formula:
C₁₀H₁₂Cl₂O₂ Melting Pt: 92…93 °C |
CAS Number:
37142-37-3 |
Specification Test Results
Melting point | 92-93°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...