Methyl-2-{4-[(tert-butoxycarbonyl)amino]piperidino}-2-phenylacetate ≥95%

Supplier: Matrix Scientific

044650-500MG 044650-1G 044650-5G
101866-910EA 375.38 USD
101866-910 101866-912 101866-914
Methyl-2-{4-[(tert-butoxycarbonyl)amino]piperidino}-2-phenylacetate ≥95%
Methyl-2-{4-[(tert-butoxycarbonyl)amino]piperidino}-2-phenylacetate

Hazard: Irritant

Formula: C₁₉H₂₈N₂O₄
MW: 348.45 g/mol
Melting Pt: 104…106 °C
MDL Number: MFCD09152694
CAS Number: 952182-80-8

Order Now

Specification Test Results

Melting point 104-106°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR