Methyl-4,6-dihydroxynicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4,6-dihydroxynicotinate

044647-500MG 044647-1G 044647-5G
101866-894EA 66.15 USD
101866-894 101866-896 101866-898
Methyl-4,6-dihydroxynicotinate ≥95%
Methyl-4,6-dihydroxynicotinate

Hazard: Irritant

Formula: C₇H₇NO₄
MW: 169.14 g/mol
Melting Pt: 252…255 °C
MDL Number: MFCD09152693
CAS Number: 79398-27-9

Order Now

Specification Test Results

Melting point 252-255°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR