1-(4-Chlorophenyl)-2-hydroxy-1-ethanone ≥95%

Supplier: Matrix Scientific

044642-500MG 044642-1G 044642-5G
101866-864EA 181.54 USD
101866-864 101866-866 101866-868
1-(4-Chlorophenyl)-2-hydroxy-1-ethanone ≥95%
1-(4-Chlorophenyl)-2-hydroxy-1-ethanone

Hazard: Irritant

Formula: C₈H₇ClO₂
Melting Pt: 127…129 °C
MDL Number: MFCD01570063
CAS Number: 27993-56-2

Order Now

Specification Test Results

Melting point 127-129°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR