1-(4-Bromophenyl)-2-hydroxy-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-Bromo-2-hydroxyacetophenone

044641-500MG 044641-1G 044641-5G
101866-858EA 152.31 USD
101866-858 101866-860 101866-862
1-(4-Bromophenyl)-2-hydroxy-1-ethanone ≥95%
1-(4-Bromophenyl)-2-hydroxy-1-ethanone

Hazard: Irritant

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 140…143 °C
MDL Number: MFCD00174275
CAS Number: 3343-45-1

Order Now

Specification Test Results

Melting point 140-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR