6-(4-Benzylpiperazino)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-(4-Benzylpiperazin-1-yl)pyridine-3-carbaldehyde

044635-500MG 044635-1G 044635-5G
101866-834EA 556.92 USD
101866-834 101866-836 101866-838
6-(4-Benzylpiperazino)nicotinaldehyde ≥95%
6-(4-Benzylpiperazino)nicotinaldehyde

Hazard: Irritant

Formula: C₁₇H₁₉N₃O
Melting Pt: 82…85 °C
MDL Number: MFCD04973728
CAS Number: 886360-69-6

Order Now

Specification Test Results

Melting point 82-85°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR