6-Morpholinonicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Morpholinylpyridine-3-carboxaldehyde

044634-500MG 044634-1G 044634-5G
101866-828EA 556.92 USD
101866-828 101866-830 101866-832
6-Morpholinonicotinaldehyde ≥95%
6-Morpholinonicotinaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD03659703
CAS Number: 173282-60-5

Order Now

Specification Test Results

Melting point 86-88°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR