6-(1,4-Thiazinan-4-yl)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-(Thiomorpholin-4-yl)pyridine-3-carbaldehyde

044631-500MG 044631-1G 044631-5G
101866-810EA 649.23 USD
101866-810 101866-812 101866-814
6-(1,4-Thiazinan-4-yl)nicotinaldehyde ≥95%
6-(1,4-Thiazinan-4-yl)nicotinaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂N₂OS
Melting Pt: 110…112 °C
MDL Number: MFCD04973725
CAS Number: 886360-66-3

Order Now

Specification Test Results

Melting point 110-112°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR