2-Hydroxy-5-sulfonicotinic acid ≥95%

Supplier: Matrix Scientific

044627-500MG 044627-1G 044627-5G
101866-792EA 215.38 USD
101866-792 101866-794 101866-796
2-Hydroxy-5-sulfonicotinic acid ≥95%
2-Hydroxy-5-sulfonicotinic acid

Hazard: Irritant

Formula: C₆H₅NO₆S
MW: 219.17 g/mol
Melting Pt: 273 °C
MDL Number: MFCD04973724
CAS Number: 334708-05-3

Order Now

Specification Test Results

Melting point 273°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR