2,5-Dichloro-3-formylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2,5-Dichloronicotinaldehyde, 2,5-Dichloro-3-formylpyridine
044626-1G
044626-5G
101866-788EA
66.15
USD
101866-788
101866-790
2,5-Dichloro-3-formylpyridine ≥95%
2,5-Dichloro-3-formylpyridine
Hazard: Irritant
Formula:
C₆H₃Cl₂NO MW: 176.0000 g/mol Melting Pt: 89…92 °C |
MDL Number:
MFCD08056627 CAS Number: 176433-49-1 |
Specification Test Results
Melting point | 89-92°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...