3-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-(3-thienyl)propanoic acid ≥97%

Supplier: Matrix Scientific

044623-500MG 044623-1G
101866-778EA 747.69 USD
101866-778 101866-780
3-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-(3-thienyl)propanoic acid ≥97%
3-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-(3-thienyl)propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₂S
Melting Pt: 112…115 °C
MDL Number: MFCD03848568
CAS Number: 866019-28-5

Order Now

Specification Test Results

Melting point 112-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR