3-Ethoxy-4-[(5-nitro-2-pyridinyl)oxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

044622-500MG 044622-1G 044622-5G
101866-772EA 181.54 USD
101866-772 101866-774 101866-776
3-Ethoxy-4-[(5-nitro-2-pyridinyl)oxy]benzenecarbaldehyde ≥95%
3-Ethoxy-4-[(5-nitro-2-pyridinyl)oxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₅
Melting Pt: 118…120 °C
MDL Number: MFCD03791225
CAS Number: 702646-74-0

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR