3-{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}propanoic acid ≥95%

Supplier: Matrix Scientific

044621-500MG 044621-1G 044621-5G
101866-766EA 556.92 USD
101866-766 101866-768 101866-770
3-{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}propanoic acid ≥95%
3-{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}propanoic acid

Hazard: Irritant

Formula: C₁₂H₉F₃N₂O₃
Melting Pt: 90…92 °C
MDL Number: MFCD03787716
CAS Number: 866018-88-4

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR