5-Amino-1-(3-nitropyridin-2-yl)-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

044620-500MG 044620-1G 044620-5G
101866-760EA 278.46 USD
101866-760 101866-762 101866-764
5-Amino-1-(3-nitropyridin-2-yl)-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-(3-nitropyridin-2-yl)-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₉H₆N₆O₂
MW: 230.19 g/mol
Melting Pt: 215…217 °C
MDL Number: MFCD03787714
CAS Number: 956754-94-2

Order Now

Specification Test Results

Melting point 215-217°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR