5-Bromo-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

044619-500MG 044619-1G 044619-5G
101866-754EA 375.38 USD
101866-754 101866-756 101866-758
5-Bromo-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylic acid ≥95%
5-Bromo-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆BrClN₂O₂
Melting Pt: 196…198 °C
MDL Number: MFCD03791224
CAS Number: 959576-61-5

Order Now

Specification Test Results

Melting point 196-198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR