Ethyl-5-bromo-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

044618-500MG 044618-1G 044618-5G
101866-748EA 278.46 USD
101866-748 101866-750 101866-752
Ethyl-5-bromo-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylate ≥95%
Ethyl-5-bromo-1-(3-chlorophenyl)-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀BrClN₂O₂
Melting Pt: 73…74 °C
MDL Number: MFCD03791223
CAS Number: 110821-39-1

Order Now

Specification Test Results

Melting point 73-74°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR