1-(2,4-Dichlorophenyl)-1H-pyrazol-4-ol ≥95%

Supplier: Matrix Scientific

044614-500MG 044614-1G 044614-5G
101866-724EA 375.38 USD
101866-724 101866-726 101866-728
1-(2,4-Dichlorophenyl)-1H-pyrazol-4-ol ≥95%
1-(2,4-Dichlorophenyl)-1H-pyrazol-4-ol

Hazard: Irritant

Formula: C₉H₆Cl₂N₂O
Melting Pt: 138…140 °C
MDL Number: MFCD03791221
CAS Number: 77458-39-0

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR