{4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanamine ≥95%

Supplier: Matrix Scientific

044612-500MG 044612-1G 044612-5G
101866-712EA 1109.23 USD
101866-712 101866-714 101866-716
{4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanamine ≥95%
{4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanamine

Hazard: Irritant

Formula: C₁₂H₁₁F₃N₂S
Melting Pt: 81…83 °C
MDL Number: MFCD03791219
CAS Number: 690632-25-8

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR