Ethyl-1-(4-fluorophenyl)-5-iodo-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

044611-500MG 044611-1G 044611-5G
101866-706EA 278.46 USD
101866-706 101866-708 101866-710
Ethyl-1-(4-fluorophenyl)-5-iodo-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-(4-fluorophenyl)-5-iodo-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀FIN₂O₂
MW: 360.13 g/mol
Melting Pt: 114…116 °C
MDL Number: MFCD03791218
CAS Number: 959576-71-7

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR