4-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

044608-500MG 044608-1G 044608-5G
101866-694EA 181.54 USD
101866-694 101866-696 101866-698
4-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid ≥95%
4-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₃
MW: 258.28 g/mol
Melting Pt: >300 °C
MDL Number: MFCD03848524
CAS Number: 866018-45-3

Order Now

Specification Test Results

Melting point 300+°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR