5-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

044606-500MG 044606-1G 044606-5G
101866-682EA 120 USD
101866-682 101866-684 101866-686
5-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid ≥95%
5-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₅F₃O₂S
MW: 246.21 g/mol
Melting Pt: 192…194 °C
MDL Number: MFCD03012933
CAS Number: 244126-64-5

Order Now

Specification Test Results

Melting point 192-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR