Methyl-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

044605-500MG 044605-1G 044605-5G
101866-676EA 270.77 USD
101866-676 101866-678 101866-680
Methyl-5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥95%
Methyl 5-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₇F₃O₂S
MW: 260.2323 g/mol
Melting Pt: 123-124°C
MDL Number: MFCD03012932
CAS Number: 146137-92-0

Order Now

Specification Test Results

Melting point 123-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR